missing translation for 'onlineSavingsMsg'
Learn More

NLK Rabbit anti-Human, Mouse, Rat, Clone: 6G5V10, Novus Biologicals™

Rabbit Monoclonal Antibody

1815.00 NOK - 4580.00 NOK

Specifications

Antigen NLK
Clone 6G5V10
Dilution Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18367335
View Documents
Bio-Techne
NBP3-15728-20UL
20 μg
1815.00 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
18329551
View Documents
Bio-Techne
NBP3-15728-100UL
100 μg
4580.00 NOK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NLK Monoclonal antibody specifically detects NLK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

NLK
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
DKFZp761G1211, EC 2.7.11, EC 2.7.11.24, FLJ21033, LAK1, nemo like kinase, nemo-like kinaseserine/threonine-protein kinase NLK, Protein LAK1
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human NLK (Q9UBE8). AVHLLCRMLVFDPSKRISAKDALAHPYLDEGRLRYHTCMCKCCFSTSTGRVYTSDFEPVTNPKFDDTFEKNLSSVRQVKEIIHQFILEQQKGNRVPLCINP
Primary
Store at -20°C. Avoid freeze-thaw cycles.
6G5V10
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Protein Kinase, Wnt Signaling Pathway
PBS, 0.05% BSA, 50% glycerol, pH7.3
51701
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.