Learn More
Nitrite, Quant™ Test Strips
Brand:  Thermo Scientific Alfa Aesar 023283.QA
Product Code. 15411137
  • 2420.00 NOK / Each
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Measures 1 to 50ppm NO2- in aqueous solutio
Nitrite test strip
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ12C004 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number023283.QA
Lot NumberA13X016 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number023283.QA
Lot NumberF09X064 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number023283.QA
Lot NumberG11X064 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number023283.QA
Product Title
Nitrite, Quant™ Test Strips > 1kit