missing translation for 'onlineSavingsMsg'
Learn More
Nitric Acid, 65% w/w, Pharmpur™, Scharlab Product Code.: 11392455
compliance-icons compliance-icons

Nitric Acid, 65% w/w, Pharmpur™, Scharlab

Product Code. 11392455
1 L
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11392455

Brand: Scharlab AC16011000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Suitable for determinations with dithizone. Scharlab Nitric Acid, 65% w/w, Pharmpur™ is also used as an oxidizing agent, for the synthesis of nitrates and organic nitro compounds, and in the pharmaceutical industry.

  • Molecular Formula: HNO3
  • Molar Mass: 63.01g/mol
  • CAS: 7697-37-2
  • EIN ECS No.: 231-714-2

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Melting Point ∼-32°C
Boiling Point 122°C
Density 1.41g/cm3
Residue after Ignition 0.0003% max
Vapor Pressure 9.4hPa at 20°C
Quantity 1 L
Physical Form Liquid
Solubility Information Miscible in water (20°C)
Formula Weight 63.01g/mol
Percent Purity ≥65% (acidimetric)
Grade Pharmpur™
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nitric acid, solution min. 65% w/w, Pharmpur, ISO, Reag. Ph Eur, for determinations with dithizone
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : nitric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nitric Acid, 65% w/w, Pharmpur™, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.