Learn More
Nitazoxanide-d4, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Nitazoxanide D4 (NTZ D4) is the deuterium labeled Nitazoxanide, which is an antiprotozoal agent.
Specifications
Specifications
Chemical Name or Material | Nitazoxanide-d4 |
CAS | 1246819-17-9 |
Purity Grade Notes | Research |
Physical Form | Solid |
Molecular Formula | C12H5D4N3O5S |
Color | Off-White |
Synonym | NTZ-d4 NSC-697855-d4 |
SMILES | O=C(NC1=NC=C([N+]([O-])=O)S1)C2=C(OC(C)=O)C([2H])=C([2H])C([2H])=C2[2H] |
Molecular Weight (g/mol) | 311.31 |
Quantity | 5 mg |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Nitazoxanide-d4, MedChemExpress