missing translation for 'onlineSavingsMsg'
Learn More
Nigrosin, soluble in spirit, Thermo Scientific™ Product Code.: 11459397

Nigrosin, soluble in spirit, Thermo Scientific™

Product Code. p-7025763
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11459397

Brand: Thermo Scientific Alfa Aesar B21424.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11099-03-9
Molecular Formula C8H19ClN2O
Molecular Weight (g/mol) 194.703
MDL Number MFCD00149005
InChI Key GBPWTMZRCAESKB-UHFFFAOYSA-N
Synonym 2-amino-n,n-dipropylacetamide hydrochloride, solvent black 5, nigrosine spirit soluble solvent black 5 and its salts, when used as a substance in hair dye products
PubChem CID 19828004
IUPAC Name 2-amino-N,N-dipropylacetamide;hydrochloride
SMILES CCCN(CCC)C(=O)CN.Cl

Specifications

Melting Point >300°C
Quantity 25 g
Solubility Information Soluble in spirit
Chemical Name or Material Nigrosin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10137087 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Lot Number10200621 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Lot Number10208600 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Lot Number10209140 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code
Lot Number10196619 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nigrosin, soluble in spirit, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.