Learn More
R&L Enterprises™ Hoffman Clips
An excellent option for sealing flexible tubing in the laboratory. R&L Enterprises™ Hoffman Clips are made of nickel plated brass to prevent corrosion.
Brand:  R&L Enterprises™ 9054
 View more versions of this product
Product Code. 12764396
  • 1175.00 NOK / Pack of 10
Only null left
Estimated Shipment: 28-07-2025
Add to basket
Description
Description
  • Either side or bottom opening
  • Available for a range of tube thicknesses
  • Threaded bar with thumbscrew
Specifications
Specifications
Nickel-plated
For use with 40mm O.D. tubing
10 Pack
Clip
Certificates
Promotions
Promotions
Product Content Correction
Product Title
R&L Enterprises™ Hoffman Clips > For Use With: For use with 40mm O.D. tubing