Learn More
Usbeck Laborgeraete™ Nickel Plated Brass Cork Borer Sets
1190.13 NOK - 2001.00 NOK
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | ||||||
10008110
|
Usbeck Laborgeraete™
5325 |
23.75, 25.00, 26.25mm |
2001.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
10150351
|
Usbeck Laborgeraete™
5323 |
16.25, 17.50, 18.75mm |
1190.13 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
Specifications
Bork Borer Set | |
Nickel Plated Brass |
Product Title
Usbeck Laborgeraete™ Nickel Plated Brass Cork Borer Sets