Learn More
Usbeck Laborgeraete™ Nickel Double Clamp Sleeves
Use Usbeck Laborgeraete™ Nickel Double Clamp Sleeves to apply 360 degrees of uniform clamping pressure for perfect fit-ups. The sleeves feature a high strength modular design for safer handling.
Brand:  Usbeck Laborgeraete™ 2201
 View more versions of this product
Product Code. 10140081
  • 85.51 NOK / Each
Only null left
Estimated Shipment: 05-08-2025
Description
  • Zinc diecasting
  • For rods up to 16 mm Ø
  • Use with nickel-plated steel or brass screws
Specifications
Specifications
1 Ea.
Double Sleeve
Nickel, Powder-coated Zinc
Nickel plated brass screws 111 g
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Usbeck Laborgeraete™ Nickel Double Clamp Sleeves > Nickel Plated Brass; Coating: Powder