Learn More
Usbeck Laborgeraete™ Nickel Double Clamp Sleeves
Use Usbeck Laborgeraete™ Nickel Double Clamp Sleeves to apply 360 degrees of uniform clamping pressure for perfect fit-ups. The sleeves feature a high strength modular design for safer handling.
Brand:  Usbeck Laborgeraete™ 2203
 View more versions of this product
Product Code. 10126821
  • 131.00 NOK / Each
Only null left
Estimated Shipment: 01-09-2025
Description
  • Zinc diecasting
  • For rods up to 16 mm Ø
  • Use with nickel-plated steel or brass screws
Specifications
Specifications
1 Ea.
Double Sleeve
Nickel, Zinc
Nickel plated brass screws 111 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Usbeck Laborgeraete™ Nickel Double Clamp Sleeves > Nickel Plated Brass