Learn More
R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems FCSTM21-30
 View more versions of this product
Product Code. 30002204
  • 61285.00 NOK / Each
Explore more special offers
Specifications
30 Plex
Multiplex Cytokine Assay Kits
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Primate
ELISA
Product Title
R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit > 30 Plex