Learn More
R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand: R&D Systems FCSTM21-10
This item is not returnable.
View return policy
Description
Specifications
10 Plex | |
Multiplex Cytokine Assay Kits | |
Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
Primate | |
ELISA |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit > 10 Plex