Learn More
R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems FCSTM21-40
 View more versions of this product
Product Code. 18441820
  • 77110.00 NOK / 1 set
Explore more special offers
Specifications
40 Plex
Multiplex Cytokine Assay Kits
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Primate
ELISA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit > 40 Plex