missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27190-1-AP-150UL
This item is not returnable.
View return policy
Description
NHE3 is an epithelial brush border Na/H exchanger that uses an inward sodium ion gradient to expel acids from the cell. NHE3 is involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. Further, NHE3 is a major proton extruding system driven by the inward sodium ion chemical gradient. NHE3 plays an important role in signal transduction. Defects in NHE3 are a cause of congenital secretory sodium diarrhea. Pseudogenes of NHE3 exist on chromosomes 10 and 22. Diseases associated with SLC9A3 include Diarrhea 8, Secretory Sodium, Congenital and Diarrhea.Specifications
NHE3 | |
Polyclonal | |
Unconjugated | |
SLC9A3 | |
Na(+)/H(+) exchanger 3, NHE 3, NHE3, SLC9A3, Sodium/hydrogen exchanger 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
105243, 6550 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
G3X939, P48764 | |
SLC9A3 | |
NHE3 Fusion Protein Ag25637 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction