missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals NG-Nitro-L-arginine-methyleester hydrochloride, 98%
1508.23 NOK - 7218.76 NOK
Chemical Identifiers
CAS | 51298-62-5 |
---|---|
Molecular Formula | C7H16ClN5O4 |
Molecular Weight (g/mol) | 269.69 |
MDL Number | MFCD00039052,MFCD00133613 |
InChI Key | QBNXAGZYLSRPJK-JEDNCBNOSA-N |
Synonym | h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome |
PubChem CID | 135193 |
IUPAC Name | methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride |
SMILES | [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10275621
|
Thermo Scientific Chemicals
328750050 |
5 g | Glass Bottle |
1508.23 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10015733
|
Thermo Scientific Chemicals
328750250 |
25 g | Glass Bottle |
7218.76 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51298-62-5 | |
269.69 | |
QBNXAGZYLSRPJK-JEDNCBNOSA-N | |
135193 | |
[H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O |
C7H16ClN5O4 | |
MFCD00039052,MFCD00133613 | |
h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome | |
methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride |
Specifications
51298-62-5 | |
100.0 | |
98% | |
Authentic | |
h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome | |
[H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O | |
+14.0° to +16.0° (24°C, 589nm) (c=3, CH3OH) | |
135193 | |
≥97.5% | |
Crystalline Solid |
97.5 | |
White | |
C7H16ClN5O4 | |
MFCD00039052,MFCD00133613 | |
QBNXAGZYLSRPJK-JEDNCBNOSA-N | |
methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride | |
269.69 | |
269.7 | |
+ 14.50 (20.00°C c=3,CH3OH) | |
N-ω-Nitro-L-arginine-methyl ester hydrochloride |
RUO – Research Use Only