Learn More
LNI Swissgas NG EOLO XL 5L
PSA nitrogen generator.
Brand:  LNI Swissgas 6920.72.005
Product Code. 17802079
  • 105050.00 NOK / Each
Description
  • Needs source of dry, oil free air
Specifications
Specifications
100/115/220/230 V
Nitrogen Gas Generator
Documents
Brochures
Certificates
Promotions
Promotions
Product Content Correction
Product Title
LNI Swissgas NG EOLO XL 5L > LNI Swissgas Nitrogen Generator