missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFATC3 Mouse anti-Human, Unconjugated, Clone: 1C1H8, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | NFATC3 |
---|---|
Clone | 1C1H8 |
Concentration | 1000 μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849225
|
Proteintech
66924-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16839225
|
Proteintech
66924-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NFATC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NFATC3. This antibody recognizes human antigen. The NFATC3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
NFATC3 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
4775 | |
Primary |
1C1H8 | |
Western Blot | |
Unconjugated | |
Human | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title