Learn More
Decon™ Neutracon
Decon Neutracon, Quantity: 5000mL, Amber, Format: Liquid, pH Range: 7.5, Viscosity: 55.0cps at 23 deg.C
Brand:  Decon™ NEU5
 View more versions of this product
Product Code. 10253710
  • 779.00 NOK / 5 L
Edge
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Description
Description
Decon Neutracon, Quantity: 5000mL, Amber, Format: Liquid, pH Range: 7.5, Viscosity: 55.0cps at 23 deg.C
Biodegradable, phosphate, enzyme and chlorine bleach free, near-neutral liquid surface-active agent. Completely soluble in water and will not deposit calcium and magnesium or precipitate iron. Solutions are completely stable in air and do not absorb CO&superscript_2;. May be used in conjunction with water soluble solvents. A 13% solution is sufficient for the majority of cleaning applications. Heating the solution, or use in an ultrasonic cleaning bath will greatly reduce the cleaning time. Specifically formulated for cleaning/decontamination of non-ferrous metals, 'soft' silica glass, polymer surfaces and other materials which may be corroded, etched, degraded, discolored or weakened by acidic or alkaline cleaning agents.
Specifications
Specifications
Liquid
Amber
55.0cps at 23°C
7.5
Phosphate, Enzyme, Chlorine Bleach Free, Water
5000 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number76813 Certificate TypeCertificate of Analysis Date08/06/2021 Catalog NumberNEU5
Lot Number76773 Certificate TypeCertificate of Analysis Date08/06/2021 Catalog NumberNEU5
Promotions
Promotions
Product Content Correction
Product Title
Decon™ Neutracon > Quantity: 5000mL