Learn More
Thermo Scientific Chemicals Neurotensin, Guinea pig
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Species | Guinea Pig |
Purification Method | HPLC |
Content And Storage | -30°C to -10°C |
Format | Solid |
Molecular Weight (g/mol) | 1662.91 |
Product Type | Neurotensin, guinea pig |
Quantity | 25 mg |
Regulatory Status | RUO - research use only |
Sequence | Glp-Leu-Tyr-Glu-Asn-Lys-Ser-Arg-Arg-Pro-Tyr-Ile-Leu |
Product Title
Thermo Scientific Chemicals Neurotensin, Guinea pig >