Learn More
Hamilton™ Neuros™ 7000 Series Modified Microliter Syringes with Knurled Hub Needle
An industry first, the Neuros accurately dispenses 50nL to 100ÎĽL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
Brand: Hamilton™ 65457-01
Description
- Needle rigidity improves insertion path accuracy
- Minimal tissue damage reduces injection variability
- Reduced sample loss saves money and materials
- Fine gauge needle creates smaller injection sites
- Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Specifications
Autoclavable | |
32 G | |
0.004 in. | |
0.009 in. | |
0.103 mm | |
0.229 mm | |
Neuros™ adapter, two adjustable needle sleeves (one with a blind stop and one without) | |
1 in. | |
Microliter | |
239°F | |
50°F | |
3 | |
Ea. | |
Blunt |
Borosilicate Glass (Barrel), Tungsten Wire (Plunger) | |
0.004 in. | |
0.10 mm | |
0.308 in. | |
7.82 mm | |
Controlled animal injections | |
1 in. | |
0 to 20 mm | |
6000 psig (413.7 bar) | |
115°C | |
7000.5 KH | |
10°C | |
0.5 ÎĽL | |
Knurled Hub |