Learn More
Brand™ Counting Chambers, Improved Neubauer, Bright-Line, BLAUBRAND™, Double ruling
BLAUBRAND™ quality. Outstanding measurement precision. 100% certified quality.
Brand:  Brand™ 717810
1156.35 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Product Code. 10350141
  • 1460.00 NOK / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
Estimated Shipment: 10-07-2025
Description
  • Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.
  • Same ruling as Improved Neubauer, but with rhodium coated chamber bottom.
  • Rulings are engraved into the rhodium layer and appear bright under normal microscope settings.
  • By altering the contrast, the microscope image can be reversed, so that the rulings appear brighter or darker, as required.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Counting Device
Rhodium Coated
0.1mm
Without spring clips
1Each
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Days
    18
    Hours
    1
    Minutes
    17
    Seconds
    3
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Brand™ Counting Chambers, Improved Neubauer, Bright-Line, BLAUBRAND™, Double ruling > Without Spring Clips