Learn More
Nepafenac-d5, MedChemExpress
MedChemExpress Nepafenac D5 (AHR-9434 D5) is the deuterium labeled Nepafenac, which is a selective COX-2 inhibitor.
Brand:  Medchem Express HY-17357S
Encompass_Preferred
 View more versions of this product
Product Code. 16469807
  • 11898.14 NOK / 5mg
Explore more special offers
Description
Description
Nepafenac D5 (AHR-9434 D5) is the deuterium labeled Nepafenac, which is a selective COX-2 inhibitor.
Specifications
Specifications
Nepafenac-d5
5 mg
Research
AHR-9434-d5 AL-6515-d5
O=C(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])C2=CC=CC(CC(N)=O)=C2N
259.31
Research
1246814-53-8
Yellow
C15H9D5N2O2
DMSO : ≥ 100 mg/mL (385.64 mM)
259.31
98.0%
Solid