Learn More
Dr.Weigert™ Neodisher FA
Neodisher LaboClean FA - Alkaline detergent, liquid concentrate for the automated cleaning of surgical thermo-stable and thermolabile instruments and others medicals or laboratories utensils. Versatile and concentrated detergent.
Brand: Dr.Weigert™ 410126
This item is not returnable.
View return policy
Description
- Alkaline detergent, liquid concentrate
- neodisher LaboClean FA is used for cleaning in washer disinfection
- Free of surfactants and active chlorine Excellent material compatibility
- Cleans blood, protein, culture media and gelatine
- Dosage between 2-4 mL
Specifications
Neodisher Detergent | |
11.6 to 11.8 | |
Disodium metasilicate, potassium silicate, potassium hydoxide | |
Canister | |
390 x 290 x 240 mm | |
25 kg |
Liquid | |
Non-sterile | |
1 to 4 mL/L | |
1.5 g/cm3 (20°C) | |
Filtered |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- neodisher FA
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : disodium metasilicate, dipotassium metasilicate, potassium hydroxide