missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK2 Rabbit anti-Human, Clone: S03-3D8, Novus Biologicals™
Rabbit Monoclonal Antibody
2690.00 NOK - 5435.00 NOK
Specifications
Antigen | NEK2 |
---|---|
Clone | S03-3D8 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18329362
|
Bio-Techne
NBP3-15065-25UL |
25 μg |
2690.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18304325
|
Bio-Techne
NBP3-15065-100UL |
100 μg |
5435.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NEK2 Monoclonal antibody specifically detects NEK2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NEK2 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
4751 | |
IgG | |
Affinity-purified |
S03-3D8 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.11, EC 2.7.11.1, HSPK 21, NEK2AHsPK21, Never in mitosis A-related kinase 2, NIMA (never in mitosis gene a)-related kinase 2, NimA-like protein kinase 1, NimA-related protein kinase 2, NLK1serine/threonine-protein kinase Nek2 | |
A synthetic peptide of human NEK2 (Uniprot#: P51955) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title