missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Needle L-Shaped Platinum Electrode
Ultra thin diameter Needle L-Shaped Platinum Electrode for in vivo applications.
Brand: BTX™ 450509
This item is not returnable.
View return policy
Description
- Enables pinpoint transfection for greater ease and efficiency in fragile tissue
- L-shaped electrodes available in 3 mm tip length
Specifications
Ex-Vivo or in-Vivo Tissues Gene or Drug Delivery, Nuclear Transfer | |
Platinum | |
Electroporation accessory | |
Needle L-Shaped Platinum Electrode, 3 mm | |
Electrode Only | |
3 mm | |
Non-sterile | |
Digital User |
Gemini X2, ECM 830, ECM 2001+ and Legacy ECM 2001 | |
BTX Electroporation | |
100 VDC | |
L-Shaped | |
0.12 in. | |
10 μsec. to 100 msec. | |
Banana Cables | |
Square Wave |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BTX™ Needle L-Shaped Platinum Electrode > L-Shaped Needle Platinum Electrode; Length: 3 mm (0.12 in.)
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Spot an opportunity for improvement?Share a Content Correction