Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Brand:  Thermo Scientific™ C4012-102SS
8852.76 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15267298
  • 8900.00 NOK / Each
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.
Crimpers provide a reproducible, secure vial closure; decrimpers allow easy removal of aluminum seals without breakage.
  • Stainless steel construction
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
Aluminum Seal
Stainless Steel
National
Manual Decrimper
11 mm
Industrial Chromatography
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms > Decrimper, for 11mm vial closure