Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Brand:  Thermo Scientific™ C4013-102SS
8469.93 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15210379
  • 8515.00 NOK / Each
Description
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.
Crimpers provide a reproducible, secure vial closure; decrimpers allow easy removal of aluminum seals without breakage.
  • Stainless steel construction
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
Aluminum Seal
Industrial Chromatography
Manual Decrimper
13 mm
National
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms > Decrimper, for 13mm vial clousre