Learn More
Thermo Scientific™ National Inserts for Standard-Opening Vials
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
Brand: Thermo Scientific™ C4012-K530
Description
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
- Pulled point inserts are an economical choice for non-critical applications
Specifications
C4012-K530 | |
200 μL | |
Conical | |
Insert |
Kimshield Glass | |
Polyspring, Pulled point | |
National |