Learn More
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Click to view available options
Color:
Amber
Clear
Closure Type:
Phenolic
Polypropylene
Polypropylene, Flanged
Septum:
PTFE
PTFE/Silicone
PTFE/Silicone, Pre-slit
PTFE/Silicone/PTFE
Packaging Type:
Polybag
Storage Tray
Unit Size:
Pack of 100
Description
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
- Packaged in convenient vial trays with clear covers or in economical polybags
- Vial trays are stackable for easy storage
- All caps are manufactured from virgin polypropylene unless noted
Includes:
100 vials with pre-attached caps and septa
Specifications
Specifications
| Closure Color | Black |
| Color (Seal) | Red/White |
| Description | Amber Glass; PP Black Cap; Red PTFE/White Silicone septum; in polybag |
| Color | Amber |
| Material | Glass, PTFE, Silicone |
| Volume (Metric) | 2 mL |
| Septum | PTFE/Silicone |
| Packaging Type | Polybag |
| Closure Material | Polypropylene |
| Closure Type | Polypropylene |
| Show More |