Learn More
Naspm trihydrochloride, MedChemExpress
MedChemExpress Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.
Brand:  Medchem Express HY-12506A
Encompass_Preferred
 View more versions of this product
Product Code. 16405906
  • 14539.45 NOK / 100mg
Explore more special offers
Description
Description
Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.
Specifications
Specifications
Naspm trihydrochloride
1049731-36-3
100 mg
C22H37Cl3N4O
1-Naphthylacetyl spermine trihydrochloride
[H]Cl.[H]Cl.[H]Cl.O=C(NCCCNCCCCNCCCN)CC1=C2C=CC=CC2=CC=C1
479.91
Research
Solid
Pale Yellow
Research
Neuroscience-Neuromodulation
H2O : 50 mg/mL (104.19 mM; Need ultrasonic) ∣DMSO : 6.4 mg/mL (13.34 mM; Need ultrasonic)
479.91
98.0%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Naspm trihydrochloride, MedChemExpress