Learn More
Velp Scientifica™ NaOH Tank with Cap
Velp Scientifica™ NaOH Tank with Cap for use with VELP UDK distillation units.
Brand:  Velp Scientifica™ A00000265
930.05 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15332499
  • 959.00 NOK / Each
Focus On Clean Water
Use promo code 24139 at checkout
View Available Promotions
Only null left
Estimated Shipment: 14-08-2025
Description
  • For use with VELP UDK distillation units
Specifications
Specifications
Cap
VELP UDK Distillation Units
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Shop now for up to 60% off Clean Water Research essentials
    Promo Code: 24139
    Expires2025-08-21
    New Offer
    Save Now
Product Content Correction
Product Title
Velp Scientifica™ NaOH Tank with Cap > Includes: Cap