Learn More
Thermo Scientific™ Nalgene™ Rubber Stopper, for Vacuum Manifold
Replace the parts and accessories on the Nalgene™ Vacuum Manifold with the Nalgene Vacuum Manifold Accessories.
Brand: Thermo Scientific™ DS0396-0080
496.03 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
- Compatible with Nalgene Vaccum Manifold (Cat. No. DS0345-0001)
- Rubber stopper, number 8
- 5/8 in. hole
Specifications
15.875 mm | |
Rubber | |
8 |
Nalgene Vacuum Manifold (Cat. No. DS0345-0001) | |
Rubber Stopper |