Learn More
Thermo Scientific™ Nalgene™ Stainless-Steel Forceps, bent tip
Nalgene Stainless-Steel Forceps are available in bent or straight-tip styles. Bent-tip style is designed for easy removal and manipulation of filter membranes.
Brand: Thermo Scientific™ DS0399-0001
432.02 NOK valid until 2025-07-04
Use promo code "24112" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
- Stainless steel
- Self-closing (bent tip)
- Serrated gripping area
- Autoclavable
Specifications
Bent, Round, Serrated | |
Stainless Steel | |
Stainless-Steel Forceps, bent tip | |
Self-closing Bent |
1 | |
Forceps | |
Ea. |
Promotions
-
Multiple Manufacturers
New Offer Save Now