Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Conical-Bottom Centrifuge Bottle Adapter
GREENER_CHOICE
Adapt Thermo Scientific™ Nalgene™ centrifuge bottles Cat. Nos. 3143, 3144 and 3145 for use in a conical-bottom centrifuge with this adapter.
Brand:  Thermo Scientific™ DS3126-0175
714.80 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10788851
  • 738.00 NOK / Pack of 4
Description
Adapt Thermo Scientific™ Nalgene™ centrifuge bottles Cat. Nos. 3143, 3144 and 3145 for use in a conical-bottom centrifuge with this adapter.
For use with Nalgene 3143-0175 polypropylene copolymer centrifuge bottles, 3144-0175 polycarbonate conical centrifuge bottles, and 3145-0175 polystyrene conical centrifuge bottles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter
Conical Bottom Centrifuge Bottle Adapter
4
Polycarbonate
White
Documents
Schematics & Diagrams
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1420727 Certificate TypeCertificate of Compliance Date12/02/2025 Catalog NumberDS3126-0175
Lot Number1409497 Certificate TypeCertificate of Compliance Date29/06/2024 Catalog NumberDS3126-0175
Lot Number1403590 Certificate TypeCertificate of Compliance Date20/03/2024 Catalog NumberDS3126-0175
Lot Number1401935 Certificate TypeCertificate of Compliance Date08/03/2024 Catalog NumberDS3126-0175
Lot Number1397532 Certificate TypeCertificate of Compliance Date25/01/2024 Catalog NumberDS3126-0175
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Polycarbonate Conical-Bottom Centrifuge Bottle Adapter > Conical Bottom Centrifuge Bottle Adapter