missing translation for 'onlineSavingsMsg'
Learn More
N-Phenylanthranilic acid, 99% Product Code.: 11416907

N-Phenylanthranilic acid, 99%

Product Code. 11416907
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11416907

Brand: Thermo Scientific Alfa Aesar A14340.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-40-7 | C13H11NO2 | 213.236 g/mol

DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.

Solubility
Soluble in ethanol, DMSO (50 mg/ml), hot alcohol, and methanol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-40-7
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
MDL Number MFCD00002421
InChI Key ZWJINEZUASEZBH-UHFFFAOYSA-N
Synonym n-phenylanthranilic acid, 2-phenylamino benzoic acid, fenamic acid, diphenylamine-2-carboxylic acid, phenylanthranilic acid, 2-carboxydiphenylamine, benzoic acid, 2-phenylamino, o-anilinobenzoic acid, n-phenyl-o-aminobenzoic acid, n-phenyl-2-aminobenzoic acidShow MoreShow Less
PubChem CID 4386
ChEBI CHEBI:34756
IUPAC Name 2-anilinobenzoic acid
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 185°C to 188°C
Quantity 100 g
Beilstein 1456607
Merck Index 14,7273
Solubility Information Soluble in ethanol,DMSO (50mg/ml),hot alcohol,and methanol. Insoluble in water.
Formula Weight 213.24
Percent Purity 99%
Chemical Name or Material N-Phenylanthranilic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249203 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog NumberA14340.22,A14340.14,A14340.36
Lot Number10245603 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog NumberA14340.22,A14340.14,A14340.36
Lot NumberJ1238A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14340.22,A14340.14,A14340.36
Lot Number10152363 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14340.22,A14340.14,A14340.36
Lot Number10160170 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14340.22,A14340.14,A14340.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Phenylanthranilic acid, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.