missing translation for 'onlineSavingsMsg'
Learn More
N,N'-Diisopropylcarbodiimide, MP Biomedicals™ Product Code.: 11401171

N,N'-Diisopropylcarbodiimide, MP Biomedicals™

Product Code. 11401171 Shop All MP Biomedicals Products
100g
Click to view available options
100g
This item is not returnable. View return policy

Product Code. 11401171

Brand: MP Biomedicals 0215775580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 693-13-0
Color Yellow
Density 0.806g/mL
Molecular Formula C7H14N2
Quantity 100 g
Synonym n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
InChI Key BDNKZNFMNDZQMI-UHFFFAOYSA-N
SMILES CC(C)N=C=NC(C)C
IUPAC Name N,N'-di(propan-2-yl)methanediimine
Molecular Weight (g/mol) 126.203
PubChem CID 12734
ChEBI CHEBI:53092
Formula Weight 126.2
Physical Form Liquid
Chemical Name or Material N, N'-Diisopropylcarbodiimide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N'-Diisopropylcarbodiimide, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.