missing translation for 'onlineSavingsMsg'
Learn More
N,N'-Dibenzylethylenediamine diacetate, 99% Product Code.: 11304865

N,N'-Dibenzylethylenediamine diacetate, 99%

Product Code. p-7047203
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11304865

Brand: Thermo Scientific Alfa Aesar L17974.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-75-8 | C20H28N2O4 | 360.454 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-75-8
Molecular Formula C20H28N2O4
Molecular Weight (g/mol) 360.454
MDL Number MFCD00040588
InChI Key MTRNNCLQPVCDLF-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine diacetate, n1,n2-dibenzylethane-1,2-diamine diacetate, n,n'-dibenzyl ethylenediamine diacetate, benzathine diacetate, dbed diacetate, unii-x18b8x2c22, n,n'-dibenzylethylenediammonium di acetate, 1,2-ethanediamine, n,n'-bis phenylmethyl-, diacetate, bis acetic acid ; benzathine, 1,2-di benzylamino ethane diacetateShow MoreShow Less
PubChem CID 31228
IUPAC Name acetic acid;N,N'-dibenzylethane-1,2-diamine
SMILES CC(=O)O.CC(=O)O.C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

Melting Point 117°C to 118°C
Quantity 25 g
Beilstein 3781362
Merck Index 14,1064
Formula Weight 360.46
Percent Purity 99%
Chemical Name or Material N,N'-Dibenzylethylenediamine diacetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot Number10250288 Certificate TypeCertificate of Analysis Date10/04/2024 Product CodeL17974.14,L17974.36,L17974.22
Lot Number10115421 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeL17974.14,L17974.36,L17974.22
Lot Number10042158 Certificate TypeCertificate of Analysis Date06/12/2021 Product CodeL17974.14,L17974.36,L17974.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N'-Dibenzylethylenediamine diacetate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.