missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl-4-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15443258
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23530-48-5
Molecular Formula C9H12N2O4S
Molecular Weight (g/mol) 244.265
MDL Number MFCD00458973
InChI Key IMRZQROLAZHXHR-UHFFFAOYSA-N
Synonym n-isopropyl-4-nitrobenzenesulfonamide, benzenesulfonamide, n-1-methylethyl-4-nitro, acmc-209g3r, cambridge id 5181757, 4-nitro-n-isopropylbenzenesulfonamide, n-isopropyl-4-nitro-benzenesulfonamide, 4-nitro-n-propan-2-yl benzenesulfonamide, methylethyl 4-nitrophenyl sulfonyl amine
PubChem CID 2054536
IUPAC Name 4-nitro-N-propan-2-ylbenzenesulfonamide
SMILES CC(C)NS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 244.26
Percent Purity 97%
Chemical Name or Material N-Isopropyl-4-nitrobenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'