Learn More
N-Hydroxy-2-methylpropanimidamide, 97%, Thermo Scientific™ Product Code.: 10085062
N-Hydroxy-2-methylpropanimidamide, 97%, Thermo Scientific™
Product Code. p-4510270
1 g, Amber Glass Bottle, 1g
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
Product Code. 10085062
Brand: Thermo Scientific Maybridge MO07460DA
Request Bulk or Custom Format
Chemical Identifiers
CAS 35613-84-4
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.14
MDL Number MFCD03426251,MFCD08705910,MFCD00464225
InChI Key JHRDEHLFNLLCQS-UHFFFAOYSA-N
Synonym z-n'-hydroxyisobutyrimidamide, 1z-n'-hydroxy-2-methylpropanimidamide, n-hydroxyisobutyrimidamide, n-hydroxy-2-methylpropanimidamide, z-n'-hydroxy-2-methylpropimidamide, n-hydroxyisobutyramidine, z-n'-hydroxy-2-methylpropanimidamide, isobutyramidoxime 250mg, n/'-hydroxy-2-methylpropanimidamide, propanimidamide, n-hydroxy-2-methyl
PubChem CID 9582835
IUPAC Name N'-hydroxy-2-methylpropanimidamide
SMILES CC(C)C(\N)=N/O
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 102.14
Percent Purity 97%
Chemical Name or Material N';-Hydroxy-2-methylpropanimidamide
Product Content Correction
Product Title
N-Hydroxy-2-methylpropanimidamide, 97%, Thermo Scientific™ >