Learn More
n-Dodecyl-beta-D-maltopyranoside
A non-ionic detergent useful in membrane solubilization studies | CAS: 69227-93-6 | C24H46O11 | 510.621 g/mol
Brand: Thermo Scientific Alfa Aesar J66869.06
9711.00 NOK valid until 2025-07-04
Use promo code "24108" to get your promotional price.
Quantity | 5 g |
---|
Description
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.
Solubility
Soluble in water at 10% w/v
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers
69227-93-6 | |
510.621 | |
NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
114880 | |
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triolShow More |
C24H46O11 | |
MFCD00043012 | |
n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranosideShow More | |
CHEBI:43769 | |
CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
Specifications
69227-93-6 | |
White | |
MFCD00043012 | |
55318 | |
Soluble in water at 10% w/v | |
CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O | |
510.621 | |
CHEBI:43769 | |
Powder |
224°C to 226°C | |
C24H46O11 | |
5 g | |
n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside | |
NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
114880 | |
510.62 | |
n-Dodecyl-beta-D-maltopyranoside |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P12L007 | Certificate of Analysis | 14/03/2025 | J66869.06,J66869.03,J66869.ME |
U08K027 | Certificate of Analysis | 21/08/2024 | J66869.06,J66869.03,J66869.ME |
R31K023 | Certificate of Analysis | 11/06/2024 | J66869.06,J66869.03,J66869.ME |
W20J029 | Certificate of Analysis | 06/10/2023 | J66869.06,J66869.03,J66869.ME |
S28J018 | Certificate of Analysis | 25/07/2023 | J66869.06,J66869.03,J66869.ME |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.