Learn More
n-Butyllithium, 2.2M solution in Cyclohex./Hexane, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10249972
n-Butyllithium, 2.2M solution in Cyclohex./Hexane, AcroSeal™, Thermo Scientific Chemicals
Product Code. 10249972
800 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10249972
Brand: Thermo Scientific Acros 377498000
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-72-8, 110-82-7, 110-54-3
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009414
InChI Key MZRVEZGGRBJDDB-UHFFFAOYSA-N
Synonym n-butyllithium, lithium, butyl, butyllithium, butyl lithium, n-butyl lithium, lithium butane, n-buli, buli, libu, unii-09w9a6b8zc
PubChem CID 61028
SMILES [Li]CCCC
Specifications
Color Brown to Gray
Linear Formula CH3(CH2)3Li
Density 0.7500g/mL
Solubility Information Solubility in water: vigorous reaction (forming lioh and butane).
IUPAC Name lithium;butane
Formula Weight 64.06
Percent Purity 19.5 to 20.5% (as BuLi) (Active base)
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.75
Quantity 800 mL
Flash Point −18°C
Physical Form Solution, magnesium salts may crystallize below 20°C
Chemical Name or Material n-Butyllithium, 2.2M Solution in Cyclohexane
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0409149 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number377498000,377491000
Lot NumberA0399076 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number377498000,377491000
Lot NumberA0403788 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number377498000,377491000
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Butyllithium, 2.3M (20 wt.%) solution in Cyclohexane/Hexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
n-Butyllithium, 2.2M solution in Cyclohex./Hexane, AcroSeal™, Thermo Scientific Chemicals >