Learn More
n-Butyl acrylate, 98+%, stab. with up to 50ppm 4-methoxyphenol Product Code.: 11302176
n-Butyl acrylate, 98+%, stab. with up to 50ppm 4-methoxyphenol
Product Code. 11302176
2500 mL, 2.50 L
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.50 L
500mL
Product Code. 11302176
Brand: Thermo Scientific Alfa Aesar L03476.0F
Request Bulk or Custom Format
CAS: 141-32-2 | C7H12O2 | 128.171 g/mol
n-Butyl acrylate is used in polymer manufacturing, textile, leather finishing, formulating paints and adhesive. It is also used in the production of homopolymers and co-polymers like acrylic acid and its salts, esters, amides, methacrylates, acrylonitrile, maleates, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene and unsaturated polyesters. Further, it is also used in making vitamin B1.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Butyl acrylate is used in polymer manufacturing, textile, leather finishing, formulating paints and adhesive. It is also used in the production of homopolymers and co-polymers like acrylic acid and its salts, esters, amides, methacrylates, acrylonitrile, maleates, vinyl acetate, vinyl chloride, vinylidene chloride, styrene, butadiene and unsaturated polyesters. Further, it is also used in making vitamin B1.
Solubility
Miscible with ethanol, ethyl ether and acetone. Slightly miscible with water and carbon tetrachloride.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, catalytic metals, organotin catalysts, amines, peroxides and halogens.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141-32-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00009446
InChI Key CQEYYJKEWSMYFG-UHFFFAOYSA-N
Synonym butyl acrylate, n-butyl acrylate, acrylic acid butyl ester, n-butyl propenoate, 2-propenoic acid, butyl ester, butyl 2-propenoate, butylacrylate, acrylic acid, butyl ester, acrylic acid n-butyl ester, butylester kyseliny akrylove
PubChem CID 8846
ChEBI CHEBI:3245
IUPAC Name butyl prop-2-enoate
SMILES CCCCOC(=O)C=C
Specifications
Melting Point -64°C
Density 0.895
Boiling Point 145°C to 146°C
Flash Point 39°C (102°F)
Odor Fruit-like
Linear Formula CH2=CHCO2CH2CH2CH2CH3
Refractive Index 1.418
Quantity 2500 mL
UN Number UN2348
Beilstein 1749970
Sensitivity Light sensitive
Merck Index 14,1539
Solubility Information Miscible with ethanol,ethyl ether and acetone. Slightly miscible with water and carbon tetrachloride.
Formula Weight 128.17
Percent Purity ≥98%
Chemical Name or Material n-Butyl acrylate, stabilized with up to 50ppm 4-methoxyphenol
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number10247647 Certificate TypeCertificate of Analysis Date16/06/2025 Product CodeL03476.0F,L03476.AP,L03476.AE
Lot Number10242952 Certificate TypeCertificate of Analysis Date16/04/2024 Product CodeL03476.0F,L03476.AP,L03476.AE
Lot Number10236004 Certificate TypeCertificate of Analysis Date06/03/2024 Product CodeL03476.0F,L03476.AP,L03476.AE
Lot Number10247647 Certificate TypeCertificate of Analysis Date11/08/2023 Product CodeL03476.0F,L03476.AP,L03476.AE
Lot Number10224527 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeL03476.0F,L03476.AP,L03476.AE
compliance-icons compliance-icons
Product Identifier
  • n-Butyl acrylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : n-Butyl acrylate
Product Title
n-Butyl acrylate, 98+%, stab. with up to 50ppm 4-methoxyphenol >