missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-cis-4-hydroxy-L-proline, 97%, Thermo Scientific Chemicals

Product Code. 11337449
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87691-27-8
Molecular Formula C10H17NO5
Molecular Weight (g/mol) 231.25
MDL Number MFCD02094406
InChI Key BENKAPCDIOILGV-UHFFFAOYNA-N
Synonym n-boc-cis-4-hydroxy-l-proline, 2s,4s-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, cis-1-tert-butoxycarbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s,4s-cis-1-n-boc-4-hydroxy-proline, boc-cishyp-oh, boc-cis-4-hydroxy-l-proline, cis-n-boc-4-hydroxy-l-proline, unii-h4ycj0kw0u, h4ycj0kw0u, 4s-1-tert-butoxycarbonyl-4-hydroxy-l-proline
PubChem CID 688614
IUPAC Name (2S,4S)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(O)CC1C(O)=O

Specifications

Melting Point ∽146°C (decomposition)
Quantity 1 g
Formula Weight 231.25
Percent Purity 97%
Chemical Name or Material N-Boc-cis-4-hydroxy-L-proline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'