Learn More
N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%, Thermo Scientific Chemicals
909.00 NOK - 12299.40 NOK
Chemical Identifiers
CAS | 13512-31-7 |
---|---|
Molecular Formula | C18H19NO5 |
Molecular Weight (g/mol) | 329.35 |
MDL Number | MFCD00038784 |
InChI Key | SLLMDHBKALJDBW-XISACWJONA-N |
Synonym | z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester |
PubChem CID | 7010613 |
SMILES | COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15479618
|
Thermo Scientific Alfa Aesar
H63113.06 |
5 g |
909.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15489618
|
Thermo Scientific Alfa Aesar
H63113.14 |
25 g |
3132.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15418136
|
Thermo Scientific Alfa Aesar
H63113.22 |
100 g |
12299.40 NOK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13512-31-7 | |
329.35 | |
SLLMDHBKALJDBW-XISACWJONA-N | |
7010613 |
C18H19NO5 | |
MFCD00038784 | |
z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester | |
COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1 |
Specifications
93°C to 97°C (decomposition) | |
C18H19NO5 | |
z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester | |
COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1 | |
329.35 | |
329.35 | |
Solid |
13512-31-7 | |
MFCD00038784 | |
SLLMDHBKALJDBW-XISACWJONA-N | |
methyl 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate | |
7010613 | |
98% | |
N-Benzyloxycarbonyl-L-tyrosine methyl ester |
Safety and Handling
missing translation for 'einecsNumber' : 426-320-4
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only