missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylglycinamide, 97%, Thermo Scientific Chemicals

Product Code. 11481457
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2620-63-5
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008029
InChI Key WQELDIQOHGAHEM-UHFFFAOYSA-N
Synonym n-acetylglycine amide, n-alpha-acetylglycinamide, unii-97sv9afw2z, acetamide, n-2-amino-2-oxoethyl, nalpha-acetylglycinamide, n-.alpha.-acetylglycinamide, 97sv9afw2z, ethanimidic acid, n-2-amino-2-oxoethyl, acetylglutamide, acetylglycinamide
PubChem CID 28326
IUPAC Name 2-acetamidoacetamide
SMILES CC(=O)NCC(N)=O

Specifications

Melting Point 136°C to 140°C
Odor Odorless
Quantity 1 g
Beilstein 1753982
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material N-Acetylglycinamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'