missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tryptophan, 97%, Thermo Scientific Chemicals

312.03 NOK - 2842.09 NOK

Chemical Identifiers

CAS 1218-34-4
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.27
MDL Number MFCD00065976
InChI Key DZTHIGRZJZPRDV-LBPRGKRZSA-N
Synonym n-acetyl-l-tryptophan, ac-trp-oh, acetyl-l-tryptophan, acetyltryptophan, acetyl-l-trp, ac-try, s-n-acetyltryptophan, unii-u9264t8oae, tryptophan, n-acetyl, s-2-acetamido-3-1h-indol-3-yl propanoic acid
PubChem CID 700653
ChEBI CHEBI:74640
IUPAC Name (2S)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
SMILES CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15296018
View Documents Promotion Details
Thermo Scientific Acros
451650050
5 g
312.03 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
15206028
View Documents Promotion Details
Thermo Scientific Acros
451650250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15216028
View Documents Promotion Details
Thermo Scientific Acros
451651000
100 g
2842.09 NOK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1218-34-4
246.27
DZTHIGRZJZPRDV-LBPRGKRZSA-N
700653
(2S)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
C13H14N2O3
MFCD00065976
n-acetyl-l-tryptophan, ac-trp-oh, acetyl-l-tryptophan, acetyltryptophan, acetyl-l-trp, ac-try, s-n-acetyltryptophan, unii-u9264t8oae, tryptophan, n-acetyl, s-2-acetamido-3-1h-indol-3-yl propanoic acid
CHEBI:74640
CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

1218-34-4
0.3% max.
96% min
MFCD00065976
DZTHIGRZJZPRDV-LBPRGKRZSA-N
(2S)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
700653
246.23
N-Acetyl-L-tryptophan
186.0°C to 192.0°C
Authentic
C13H14N2O3
n-acetyl-l-tryptophan, ac-trp-oh, acetyl-l-tryptophan, acetyltryptophan, acetyl-l-trp, ac-try, s-n-acetyltryptophan, unii-u9264t8oae, tryptophan, n-acetyl, s-2-acetamido-3-1h-indol-3-yl propanoic acid
CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
246.27
CHEBI:74640
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 214-935-9

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only