missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Acetyl-L-proline, 99%
639.00 NOK - 2160.00 NOK
Chemical Identifiers
CAS | 68-95-1 |
---|---|
Molecular Formula | C7H11NO3 |
Molecular Weight (g/mol) | 157.17 |
MDL Number | MFCD00020837 |
InChI Key | GNMSLDIYJOSUSW-UHFFFAOYNA-N |
Synonym | n-acetyl-l-proline, acetylproline, ac-pro-oh, n-acetylproline, 1-acetyl-l-proline, acetyl proline, l-proline, 1-acetyl, acetyl-l-proline, 2s-1-acetylpyrrolidine-2-carboxylic acid, s-1-acetylpyrrolidine-2-carboxylic acid |
PubChem CID | 66141 |
ChEBI | CHEBI:21560 |
IUPAC Name | (2S)-1-acetylpyrrolidine-2-carboxylic acid |
SMILES | CC(=O)N1CCCC1C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11393446
|
Thermo Scientific Chemicals
L14300.03 |
1 g |
639.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11303456
|
Thermo Scientific Chemicals
L14300.06 |
5 g |
2160.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68-95-1 | |
157.17 | |
GNMSLDIYJOSUSW-UHFFFAOYNA-N | |
66141 | |
(2S)-1-acetylpyrrolidine-2-carboxylic acid |
C7H11NO3 | |
MFCD00020837 | |
n-acetyl-l-proline, acetylproline, ac-pro-oh, n-acetylproline, 1-acetyl-l-proline, acetyl proline, l-proline, 1-acetyl, acetyl-l-proline, 2s-1-acetylpyrrolidine-2-carboxylic acid, s-1-acetylpyrrolidine-2-carboxylic acid | |
CHEBI:21560 | |
CC(=O)N1CCCC1C(O)=O |
Specifications
117°C to 118°C | |
C7H11NO3 | |
83200 | |
GNMSLDIYJOSUSW-UHFFFAOYNA-N | |
(2S)-1-acetylpyrrolidine-2-carboxylic acid | |
66141 | |
157.18 | |
−116° (c=1 in Water) |
68-95-1 | |
MFCD00020837 | |
n-acetyl-l-proline, acetylproline, ac-pro-oh, n-acetylproline, 1-acetyl-l-proline, acetyl proline, l-proline, 1-acetyl, acetyl-l-proline, 2s-1-acetylpyrrolidine-2-carboxylic acid, s-1-acetylpyrrolidine-2-carboxylic acid | |
CC(=O)N1CCCC1C(O)=O | |
157.17 | |
CHEBI:21560 | |
99% | |
N-Acetyl-L-proline |
Safety and Handling
missing translation for 'einecsNumber' : 200-698-9
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only