Learn More
N-Acetyl-L-isoleucine, 98%, Thermo Scientific Chemicals
Product Code. 15492819
Click to view available options
Quantity:
5 g
25 g
100 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3077-46-1
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00066072
InChI Key JDTWZSUNGHMMJM-IFSCUNPJNA-N
Synonym n-acetyl-l-isoleucine, 2s,3s-2-acetamido-3-methylpentanoic acid, ac-ile-oh, l-isoleucine, n-acetyl, unii-p9cl0y0bvz, p9cl0y0bvz, isoleucine, n-acetyl-, l, acetyl isoleucine, n-acetyl-isoleucine, acetyl-l-isoleucine
PubChem CID 7036275
ChEBI CHEBI:21555
SMILES CC[C@H](C)[C@H](NC(C)=O)C(O)=O
Specifications
Color White
IUPAC Name (2S,3S)-2-acetamido-3-methylpentanoic acid
Formula Weight 173.21
Percent Purity 98%
Quantity 100 g
Physical Form Crystalline
Chemical Name or Material N-Acetyl-L-isoleucine