Learn More
N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester, 99%, Thermo Scientific™
418.00 NOK - 6445.00 NOK
Chemical Identifiers
CAS | 13726-52-8 |
---|---|
Molecular Formula | C16H22N2O5 |
Molecular Weight (g/mol) | 322.361 |
MDL Number | MFCD00009617 |
InChI Key | RJXFBLRRPYBPTM-ZDUSSCGKSA-N |
Synonym | n-4-aminobenzoyl-l-glutamic acid diethyl ester, diethyl n-4-aminobenzoyl-l-glutamate, diethyl n-p-aminobenzoyl-l-glutamate, 1,5-diethyl 2s-2-4-aminophenyl formamido pentanedioate, 4-abz-glu oet-oet, l-glutamic acid, n-4-aminobenzoyl-, diethyl ester, diethyl 4-aminobenzoyl-l-glutamate, diethyl p-aminobenzoyl-l-glutamate, s-diethyl 2-4-aminobenzamido pentanedioate, diethyl 2-4-aminobenzoyl amino pentanedioate # |
PubChem CID | 73002 |
IUPAC Name | diethyl (2S)-2-[(4-aminobenzoyl)amino]pentanedioate |
SMILES | CCOC(=O)CCC(C(=O)OCC)NC(=O)C1=CC=C(C=C1)N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13726-52-8 | |
322.361 | |
RJXFBLRRPYBPTM-ZDUSSCGKSA-N | |
73002 | |
CCOC(=O)CCC(C(=O)OCC)NC(=O)C1=CC=C(C=C1)N |
C16H22N2O5 | |
MFCD00009617 | |
n-4-aminobenzoyl-l-glutamic acid diethyl ester, diethyl n-4-aminobenzoyl-l-glutamate, diethyl n-p-aminobenzoyl-l-glutamate, 1,5-diethyl 2s-2-4-aminophenyl formamido pentanedioate, 4-abz-glu oet-oet, l-glutamic acid, n-4-aminobenzoyl-, diethyl ester, diethyl 4-aminobenzoyl-l-glutamate, diethyl p-aminobenzoyl-l-glutamate, s-diethyl 2-4-aminobenzamido pentanedioate, diethyl 2-4-aminobenzoyl amino pentanedioate # | |
diethyl (2S)-2-[(4-aminobenzoyl)amino]pentanedioate |
Specifications
139°C to 142°C | |
C16H22N2O5 | |
n-4-aminobenzoyl-l-glutamic acid diethyl ester, diethyl n-4-aminobenzoyl-l-glutamate, diethyl n-p-aminobenzoyl-l-glutamate, 1,5-diethyl 2s-2-4-aminophenyl formamido pentanedioate, 4-abz-glu oet-oet, l-glutamic acid, n-4-aminobenzoyl-, diethyl ester, diethyl 4-aminobenzoyl-l-glutamate, diethyl p-aminobenzoyl-l-glutamate, s-diethyl 2-4-aminobenzamido pentanedioate, diethyl 2-4-aminobenzoyl amino pentanedioate # | |
CCOC(=O)CCC(C(=O)OCC)NC(=O)C1=CC=C(C=C1)N | |
322.361 | |
322.36 | |
99% |
13726-52-8 | |
MFCD00009617 | |
RJXFBLRRPYBPTM-ZDUSSCGKSA-N | |
diethyl (2S)-2-[(4-aminobenzoyl)amino]pentanedioate | |
73002 | |
19° (c=1 Chloroform) | |
N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester |
Safety and Handling
missing translation for 'einecsNumber' : 237-293-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.