missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Bromoethyl)phthalimide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 81.0°C to 84.0°C |
Color | Beige or Pink to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (on dry substance) (HPLC) |
Packaging | Glass bottle |
Quantity | 100 g |
Beilstein | 21,461 |
Solubility Information | Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents ca.10g/l,hardly soluble in ethanol and ether |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Safety and Handling
Product Identifier
- N-(2-Bromoethyl)phthalimide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-(2-Bromoethyl)phthalimide, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction