missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10906-1-AP-150UL
This item is not returnable.
View return policy
Description
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifications
Myosin Light Chain 2 | |
Polyclonal | |
Unconjugated | |
MYL2 | |
CMH10, MLC 2, MLC 2v, MLC2, MLC2V, MYL2, Myosin Light Chain 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17906, 363925, 4633, 677750 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08733, P10916, P51667 | |
Myl2, myl2b | |
Myosin Light Chain 2 Fusion Protein Ag1356 | |
150 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction